Learn More
TARGETMOL CHEMICALS INC MKK7-COV-9 5MG
SDP
Catalog No. 502579578
Encompass_Preferred
Also available in 1 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MKK7-COV-9 is a selective and potent covalent inhibitor of MKK7 with inhibitory effects on MKK7-induced protein-protein interactions.MKK7-COV-9 interrupts the activation of primary B-cells in response to LPS.MKK7-COV-9 shows low cytotoxicity at high concentrations. purity: 99%
Catalog No. 50-257-9578 Supplier TARGETMOL CHEMICALS INC Supplier No. T608575MG
Also available in 1 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MKK7-COV-9 is a selective and potent covalent inhibitor of MKK7 with inhibitory effects on MKK7-induced protein-protein interactions.MKK7-COV-9 interrupts the activation of primary B-cells in response to LPS.MKK7-COV-9 shows low cytotoxicity at high concentrations. purity: 99%