Learn More
Aviva Systems Biology Corp MKNK1 Antibody - N-terminal region (ARP60346_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60346_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MKNK1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MKNK1. Alternate names: MNK1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-014