Learn More
Eurofins Discoverx MKP5, active, 250 µg. (14-779M)
Supplier:  Eurofins Discoverx 14779M
Encompass
The MKP5, active product is purified N terminal 6xHis-tagged recombinant MKP5 enzyme. This enzyme contains amino acids 320-end. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 2.16 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9330