Learn More
Aviva Systems Biology Corp MKX Antibody - middle region (ARP37938_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37938_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MKX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Irxl1, 9430023B20RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56464