Learn More
Aviva Systems Biology Corp MKX Antibody - middle region (ARP31887_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31887_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MKX. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MKX. Alternate names: C10orf48, IFRX, IRXL1, MGC39616ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52842