Learn More
Medchemexpress LLC HY-103050 5mg Medchemexpress, ML-030 CAS:1013750-77-0 Purity:>98%
SDP
Catalog No. 501968026
Encompass_Preferred
Medchemexpress, HY-103050 5mg ML-030 CAS:1013750-77-0 ML-030 is a potent PDE4 inhibitor, with IC50 of 6.7 nM, 12.9 nM, 48.2 nM, 37.2 nM, 452 nM and 49.2 nM for PDE4A, PDE4A1, PDE4B1, PDE4B2, PDE4C1,and PDE4D2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8026 Supplier Medchemexpress LLC Supplier No. HY1030505MG
Medchemexpress, HY-103050 5mg ML-030 CAS:1013750-77-0 ML-030 is a potent PDE4 inhibitor, with IC50 of 6.7 nM, 12.9 nM, 48.2 nM, 37.2 nM, 452 nM and 49.2 nM for PDE4A, PDE4A1, PDE4B1, PDE4B2, PDE4C1,and PDE4D2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.