Learn More
TARGETMOL CHEMICALS INC ML-090 5MG
SDP
Catalog No. 502580904
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ML-090 is NOX1-specific inhibitor(IC50 = 90 nM) which is >100 selectivity for NOX1 over NOX2 NOX3 NOX4 (all IC50s >10 uM). purity: 98%
Catalog No. 50-258-0904 Supplier TARGETMOL CHEMICALS INC Supplier No. T229905MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ML-090 is NOX1-specific inhibitor(IC50 = 90 nM) which is >100 selectivity for NOX1 over NOX2 NOX3 NOX4 (all IC50s >10 uM). purity: 98%