Learn More
Cayman Chemical ML-095 (hydrochloride), Purity: ≥98%, CAS: 1135318-57-8, 10 mg
SDP
Catalog No. 502972783
Encompass_Preferred
An inhibitor of PLAP (IC50 = 3.7 μM) that demonstrates high selectivity against the related alkaline phosphatases, TNAP and IAP (IC50s > 100 μM)
Catalog No. 50-297-2783 Supplier Cayman Chemical Supplier No. 1517310 MG
An inhibitor of PLAP (IC50 = 3.7 μM) that demonstrates high selectivity against the related alkaline phosphatases, TNAP and IAP (IC50s > 100 μM)