Learn More
Medchemexpress LLC ML-099 | 496775-95-2 | 5MG
SDP
Catalog No. 5000235419
Encompass_Preferred
ML-099 (CID-888706) is a pan Ras-related GTPases activator that can activate Rac1 cell division cycle 42 Ras Rab7 and Rab-2A[1]
Catalog No. 50-002-35419 Supplier Medchemexpress LLC Supplier No. HY1243065MG
ML-099 (CID-888706) is a pan Ras-related GTPases activator that can activate Rac1 cell division cycle 42 Ras Rab7 and Rab-2A[1]