Learn More
Medchemexpress LLC ML-109 | 1186649-91-1 | 99.0% | 523.58 | 50 MG
SDP
Catalog No. 5000437341
Encompass_Preferred
ML-109 is identified as a potent and full agonist of the thyroid stimulating hormone receptor (TSHR), exhibiting an EC50 of 40 nM. While being surprisingly stable under neutral and basic conditions with a half-life of approximately 16 hours, it degrades at lower pH levels, with a half-life of about 3 hours. It shows no activity at FSHR or LHCGR.
  • Potent and full thyroid stimulating hormone receptor (TSHR) agonist
  • EC50 of 40 nM for TSHR
  • No activity at FSHR or LHCGR
  • Stable at neutral and basic conditions (t1/2 of ~16 h)
Catalog No. 50-004-37341 Supplier Medchemexpress LLC Supplier No. HY11411650MG
ML-109 is identified as a potent and full agonist of the thyroid stimulating hormone receptor (TSHR), exhibiting an EC50 of 40 nM. While being surprisingly stable under neutral and basic conditions with a half-life of approximately 16 hours, it degrades at lower pH levels, with a half-life of about 3 hours. It shows no activity at FSHR or LHCGR.
  • Potent and full thyroid stimulating hormone receptor (TSHR) agonist
  • EC50 of 40 nM for TSHR
  • No activity at FSHR or LHCGR
  • Stable at neutral and basic conditions (t1/2 of ~16 h)