Learn More
Medchemexpress LLC ML-109 | 1186649-91-1 | 99.02% | 523.58 | 100 MG
SDP
Catalog No. 5000436978
Encompass_Preferred
ML-109 is a potent and full thyroid stimulating hormone receptor (TSHR) agonist, with an EC50 of 40 nM. It is for research use only.
  • Potent and full agonist at TSHR with an EC50 of 40 nM.
  • No activity at FSHR or LHCGR.
  • Surprisingly stable under neutral and basic conditions (t1/2 of ~16 h), though it degrades at low pH (t1/2 of ~3 h).
Catalog No. 50-004-36978 Supplier Medchemexpress LLC Supplier No. HY114116100MG
ML-109 is a potent and full thyroid stimulating hormone receptor (TSHR) agonist, with an EC50 of 40 nM. It is for research use only.
  • Potent and full agonist at TSHR with an EC50 of 40 nM.
  • No activity at FSHR or LHCGR.
  • Surprisingly stable under neutral and basic conditions (t1/2 of ~16 h), though it degrades at low pH (t1/2 of ~3 h).