Learn More
Medchemexpress LLC HY-114116 5mg Medchemexpress, ML-109 CAS:1186649-91-1 Purity:>98%
SDP
Catalog No. 501969285
Encompass_Preferred
Medchemexpress, HY-114116 5mg ML-109 CAS:1186649-91-1 ML-109 is a potent and full thyroid stimulating hormone receptor (TSHR) agonist, with an EC50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9285 Supplier Medchemexpress LLC Supplier No. HY1141165MG
Medchemexpress, HY-114116 5mg ML-109 CAS:1186649-91-1 ML-109 is a potent and full thyroid stimulating hormone receptor (TSHR) agonist, with an EC50 of 40 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.