Learn More
Apexbio Technology LLC ML 120B dihydrochloride 783348-36-7 (free base) 10mg
SDP
Catalog No. 5000088532
Encompass_Preferred
ML 120B dihydrochloride is a selective agonist targeting the G-protein-coupled receptor 40 (GPR40) also known as free fatty acid receptor 1 (FFAR1) It is designed to activate GPR40 thereby modulating downstream signal transduction pathways that regulate glucose-stimulated insulin secretion in pancreatic -cells ML 120B dihydrochloride exerts its biological activity primarily through specific engagement of GPR40-mediated pathways Based on these pharmacological properties ML 120B dihydrochloride holds research potential in investigating mechanisms underlying glucose-dependent insulin secretion and in the development of therapeutic strategies for diabetes and related metabolic disorders
Catalog No. 50-000-88532 Supplier Apexbio Technology LLC Supplier No. B575610
ML 120B dihydrochloride is a selective agonist targeting the G-protein-coupled receptor 40 (GPR40) also known as free fatty acid receptor 1 (FFAR1) It is designed to activate GPR40 thereby modulating downstream signal transduction pathways that regulate glucose-stimulated insulin secretion in pancreatic -cells ML 120B dihydrochloride exerts its biological activity primarily through specific engagement of GPR40-mediated pathways Based on these pharmacological properties ML 120B dihydrochloride holds research potential in investigating mechanisms underlying glucose-dependent insulin secretion and in the development of therapeutic strategies for diabetes and related metabolic disorders