Learn More
Cayman Chemical ML-133 (hydrochloride), Purity: ≥98%, CAS: 1222781-70-5, 50 mg
SDP
Catalog No. 502991552
Encompass_Preferred
A Kir2 channel inhibitor (IC50s = 1.8, 2.8, 2,9, and 2.6 µM for Kir2.1, Kir2.2, Kir2.3, and Kir2.6, respectively); selective for Kir2 channels over Kir1.1 and Kir4.1 channels (IC50s = 33 and 76 µM, respectively); prevents the development of dynamic, but not punctate, mechanical allodynia in a mouse model of spared nerve injury at 10 and 30 nmol/animal
Catalog No. 50-299-1552 Supplier Cayman Chemical Supplier No. 3123250 MG
A Kir2 channel inhibitor (IC50s = 1.8, 2.8, 2,9, and 2.6 µM for Kir2.1, Kir2.2, Kir2.3, and Kir2.6, respectively); selective for Kir2 channels over Kir1.1 and Kir4.1 channels (IC50s = 33 and 76 µM, respectively); prevents the development of dynamic, but not punctate, mechanical allodynia in a mouse model of spared nerve injury at 10 and 30 nmol/animal