Learn More
Abcam ML-162, 5MG
Supplier:  Abcam AB2869995MG
Encompass_Preferred
MW 477.4 Da, Purity >=98%. ML-162 is a GPX4 (glutathione peroxidase 4) inhibitor that is selectively lethal to mutant RAS oncogene-expressing cell lines (IC₅₀s = 25 and 578 nM for HRASG12V-expressing and wild-type BJ fibroblasts, respectively).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5877