Learn More
Medchemexpress LLC ML-184 10mg | 794572-10-4 | 470.63 g·mol⁻¹ | C25H34N4O3S | 10 MG
SDP
Catalog No. 5000229272
Encompass_Preferred
ML-184 is a selective GPR55 agonist research compound supplied for in vitro and preclinical pharmacology studies. It is provided as a solid and as ready-to-use DMSO solutions, and is characterized by molecular formula C25H34N4O3S and molecular weight 470.63 g·mol⁻¹.
  • Selective GPR55 agonist (EC50 ≈ 250 nM)
  • Suitable for in vitro and preclinical research applications
  • Available as a 10 mg solid and as 10 mM solution in DMSO
  • High purity (99.57%) as provided by manufacturer
  • Store powder at -20°C for long-term stability; store solutions at -80°C for extended storage
Catalog No. 50-002-29272 Supplier Medchemexpress LLC Supplier No. HY11646110MG
ML-184 is a selective GPR55 agonist research compound supplied for in vitro and preclinical pharmacology studies. It is provided as a solid and as ready-to-use DMSO solutions, and is characterized by molecular formula C25H34N4O3S and molecular weight 470.63 g·mol⁻¹.
  • Selective GPR55 agonist (EC50 ≈ 250 nM)
  • Suitable for in vitro and preclinical research applications
  • Available as a 10 mg solid and as 10 mM solution in DMSO
  • High purity (99.57%) as provided by manufacturer
  • Store powder at -20°C for long-term stability; store solutions at -80°C for extended storage