Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ML-184 25mg | 794572-10-4 | 470.63 g·mol⁻¹ | C25H34N4O3S | 25 MG
SDP

Catalog No. 5000228025
Encompass_Preferred

ML-184 is a small-molecule research compound described as a GPR55 agonist and used in cell-based and pharmacological studies. It is supplied as a high-purity solid for laboratory research and is identified by CAS 794572-10-4.

  • Acts as a GPR55 agonist for pharmacology and cell studies.
  • High reported purity suitable for research applications.
  • Molecular formula C25H34N4O3S and molecular weight 470.63 g·mol⁻¹.
  • Supplied as a solid in a 25 mg package for benchtop use.
  • CAS number 794572-10-4 provides an unambiguous chemical identifier.

Catalog No. 50-002-28025 Supplier Medchemexpress LLC Supplier No. HY11646125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML-184 is a small-molecule research compound described as a GPR55 agonist and used in cell-based and pharmacological studies. It is supplied as a high-purity solid for laboratory research and is identified by CAS 794572-10-4.

  • Acts as a GPR55 agonist for pharmacology and cell studies.
  • High reported purity suitable for research applications.
  • Molecular formula C25H34N4O3S and molecular weight 470.63 g·mol⁻¹.
  • Supplied as a solid in a 25 mg package for benchtop use.
  • CAS number 794572-10-4 provides an unambiguous chemical identifier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.