Learn More
Medchemexpress LLC ML-184 50mg | 794572-10-4 | 470.63 g/mol | C25H34N4O3S | 50 MG
SDP
Catalog No. 5000307761
Encompass_Preferred
ML-184 is a selective GPR55 agonist research compound (CAS 794572-10-4) used to probe GPR55-mediated signaling pathways in vitro and in vivo. It activates ERK1/2 phosphorylation, promotes PKCβII translocation, and has been shown to increase neural stem cell proliferation and differentiation.
  • Selective GPR55 agonist with an EC50 of about 250 nM.
  • Greater than 100-fold selectivity over GPR35, CB1, and CB2.
  • Induces ERK1/2 phosphorylation and PKCβII membrane translocation.
  • Demonstrated activity in neural stem cell proliferation and differentiation assays.
  • Provided as a high-purity solid and as a DMSO solution for biological assays.
Catalog No. 50-003-07761 Supplier Medchemexpress LLC Supplier No. HY11646150MG
ML-184 is a selective GPR55 agonist research compound (CAS 794572-10-4) used to probe GPR55-mediated signaling pathways in vitro and in vivo. It activates ERK1/2 phosphorylation, promotes PKCβII translocation, and has been shown to increase neural stem cell proliferation and differentiation.
  • Selective GPR55 agonist with an EC50 of about 250 nM.
  • Greater than 100-fold selectivity over GPR35, CB1, and CB2.
  • Induces ERK1/2 phosphorylation and PKCβII membrane translocation.
  • Demonstrated activity in neural stem cell proliferation and differentiation assays.
  • Provided as a high-purity solid and as a DMSO solution for biological assays.