Learn More
Medchemexpress LLC ML-184 5mg | 794572-10-4 | 470.63 g/mol | C25H34N4O3S | 5 MG
SDP
Catalog No. 5000229304
Encompass_Preferred
ML-184 is a selective GPR55 agonist research compound (CAS 794572-10-4) used to investigate GPR55-mediated signaling. It activates GPR55 with an EC50 of approximately 250 nM, induces ERK1/2 phosphorylation and PKCβII translocation, and has been reported to increase neural stem cell proliferation and promote neuronal differentiation. The supplied material is a 5 mg vial with reported purity of 99.57% and molecular weight 470.63 g/mol.
  • Selective GPR55 agonist with EC50 ≈ 250 nM.
  • Induces ERK1/2 phosphorylation and PKCβII translocation.
  • Shown to increase neural stem cell proliferation and promote neuronal differentiation in vitro.
  • High reported purity suitable for research use.
  • Supplied as a 5 mg vial for small-scale experiments.
Catalog No. 50-002-29304 Supplier Medchemexpress LLC Supplier No. HY1164615MG
ML-184 is a selective GPR55 agonist research compound (CAS 794572-10-4) used to investigate GPR55-mediated signaling. It activates GPR55 with an EC50 of approximately 250 nM, induces ERK1/2 phosphorylation and PKCβII translocation, and has been reported to increase neural stem cell proliferation and promote neuronal differentiation. The supplied material is a 5 mg vial with reported purity of 99.57% and molecular weight 470.63 g/mol.
  • Selective GPR55 agonist with EC50 ≈ 250 nM.
  • Induces ERK1/2 phosphorylation and PKCβII translocation.
  • Shown to increase neural stem cell proliferation and promote neuronal differentiation in vitro.
  • High reported purity suitable for research use.
  • Supplied as a 5 mg vial for small-scale experiments.