Learn More
Medchemexpress LLC ML-193 | 713121-80-3 | 99.7% | 527.59 | 100 MG
SDP
Catalog No. 5000344161
Encompass_Preferred
ML-193 (CID 1261822) is a potent and selective antagonist of GPR55, with an IC50 of 221 nM. It demonstrates more than 27-fold selectivity for GPR55 over GPR35, CB1, and CB2. This compound has been shown to improve motor and sensorimotor deficits in rat models of Parkinson's disease.
  • Potent and selective GPR55 antagonist.
  • High selectivity over GPR35, CB1, and CB2.
  • Improves motor and sensorimotor deficits in Parkinson's disease rat models.
  • Available in solid and solution forms.
  • Detailed solubility information provided.
  • Includes detailed handling and storage instructions.
Catalog No. 50-003-44161 Supplier Medchemexpress LLC Supplier No. HY11012550MG
ML-193 (CID 1261822) is a potent and selective antagonist of GPR55, with an IC50 of 221 nM. It demonstrates more than 27-fold selectivity for GPR55 over GPR35, CB1, and CB2. This compound has been shown to improve motor and sensorimotor deficits in rat models of Parkinson's disease.
  • Potent and selective GPR55 antagonist.
  • High selectivity over GPR35, CB1, and CB2.
  • Improves motor and sensorimotor deficits in Parkinson's disease rat models.
  • Available in solid and solution forms.
  • Detailed solubility information provided.
  • Includes detailed handling and storage instructions.