Learn More
Abcam ML 204, TRPC4 blocker, 5MG
Catalog No. 502672002
Encompass_Preferred
MW 226.32 Da. Selective TRPC4 channel blocker (IC₅₀ = 0.96 (fluorescence assay) - 2.6 μM (electrophysiological assay)). Selective versus TRPC6 (19-fold) and TRPC5 (9-fold). Essentially inactive at TRPV1, TRPV3, TRPA1 and TRPM8 channels at concentrations up to 22 μM.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2002 Supplier Abcam Supplier No. AB2698835MG
MW 226.32 Da. Selective TRPC4 channel blocker (IC₅₀ = 0.96 (fluorescence assay) - 2.6 μM (electrophysiological assay)). Selective versus TRPC6 (19-fold) and TRPC5 (9-fold). Essentially inactive at TRPV1, TRPV3, TRPA1 and TRPM8 channels at concentrations up to 22 μM.
The product is subject to the following: Abcam Restricted Use Statement