Learn More
TARGETMOL CHEMICALS INC ML-210 10MG
SDP
Catalog No. 502579084
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-210 (CID 49766530) is a selective RAS and covalent glutathione peroxidase 4 (GPX4) inhibitor(GPX4 EC50 30 nM) with anti-cancer activitypurity: 97%
Catalog No. 50-257-9084 Supplier TARGETMOL CHEMICALS INC Supplier No. T837510MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-210 (CID 49766530) is a selective RAS and covalent glutathione peroxidase 4 (GPX4) inhibitor(GPX4 EC50 30 nM) with anti-cancer activitypurity: 97%