Learn More
Medchemexpress LLC HY-100003 5mg Medchemexpress, ML-210 CAS:1360705-96-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1000035MG
Encompass_Preferred
Medchemexpress, HY-100003 5mg ML-210 CAS:1360705-96-9 ML-210 is a selective and covalent glutathione peroxidase 4 (GPX4) inhibitor with an EC50 of 30 nM. ML-210 binds the GPX4 selenocysteine residue. ML-210 has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7707