Learn More
Cayman Chemical ML-241hydrochlorIde 10mg
SDP
Catalog No. 502942324
Encompass_Preferred
A selective inhibitor of p97 (IC50 = 0.11 µM); selective for the p97 D2 domain; disrupts the ERAD pathway, preventing the degradation of p97-dependent proteasome substrates in a reporter assay using HeLa cells (IC50 = 3.5 µM) and inducing the accumulation of ubiquitin conjugates in the nuclear plus membrane and cytosolic compartments of SW403 cells at 5 and 10 µM
Catalog No. 50-294-2324 Supplier Cayman Chemical Supplier No. 4040010 MG
A selective inhibitor of p97 (IC50 = 0.11 µM); selective for the p97 D2 domain; disrupts the ERAD pathway, preventing the degradation of p97-dependent proteasome substrates in a reporter assay using HeLa cells (IC50 = 3.5 µM) and inducing the accumulation of ubiquitin conjugates in the nuclear plus membrane and cytosolic compartments of SW403 cells at 5 and 10 µM