Learn More
Cayman Chemical ML-252 (hydrochloride), Purity: ≥98%, CAS: 2309887-61-2, 10 mg
SDP
Catalog No. 502972814
Encompass_Preferred
A potent and selective inhibitor of Kv7.2 channels (IC50 = 69 nM); more potent than the (R)-enantiomer and racemic mixture (IC50s = 944 and 160 nM, respectively); selective for Kv7.2 over other Kv channel subtypes, G protein-coupled receptors, transporters, calcium channels, and KATP and hERG potassium channels; inhibits the MT1 receptor 61% when used at a concentration of 10 µM
Catalog No. 50-297-2814 Supplier Cayman Chemical Supplier No. 1519110 MG
A potent and selective inhibitor of Kv7.2 channels (IC50 = 69 nM); more potent than the (R)-enantiomer and racemic mixture (IC50s = 944 and 160 nM, respectively); selective for Kv7.2 over other Kv channel subtypes, G protein-coupled receptors, transporters, calcium channels, and KATP and hERG potassium channels; inhibits the MT1 receptor 61% when used at a concentration of 10 µM