Learn More
Medchemexpress LLC HY-112606 5mg Medchemexpress, ML-290 CAS:1482500-76-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1126065MG
Encompass_Preferred
Medchemexpress, HY-112606 5mg ML-290 CAS:1482500-76-4 ML-290 is a first-in-class and potent relaxin/insulin-like family peptide receptor ( RXFP1 ) agonist and activator of anti-fibrotic genes, with an EC 50 of 94 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2425