Learn More
TARGETMOL CHEMICALS INC ML 297 10MG
SDP
Catalog No. 502576738
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML 297 is a selective Kir3.1/3.2 (GIRK1/2) channel activator (IC50 values are 160 887 and 914 nM for GIRK1/2 GIRK1/4 and GIRK1/3 respectively). ML 297 exhibits no effect on GIRK2 GIRK2/3 Kir2.1 and Kv7.4 channels and has minimal effect on a panel of other ion channels receptors and transporters. purity: 96%
Catalog No. 50-257-6738 Supplier TARGETMOL CHEMICALS INC Supplier No. T466410MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML 297 is a selective Kir3.1/3.2 (GIRK1/2) channel activator (IC50 values are 160 887 and 914 nM for GIRK1/2 GIRK1/4 and GIRK1/3 respectively). ML 297 exhibits no effect on GIRK2 GIRK2/3 Kir2.1 and Kv7.4 channels and has minimal effect on a panel of other ion channels receptors and transporters. purity: 96%