Learn More
Medchemexpress LLC HY-110192 10mg Medchemexpress, ML 297 CAS:1443246-62-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11019210MG
Encompass_Preferred
Medchemexpress, HY-110192 10mg ML 297 CAS:1443246-62-5 ML 297 (VU 0456810) is a potent and selective GIRK1/2 activator, with an EC50 of 0.16 μM. ML 297 is potential for the treatment of epilepsy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8765