Learn More
Selleck Chemical LLC ML-297
SDP
Supplier:  Selleck Chemical LLC S65595MG
Encompass_Preferred
ML-297 (VU 0456810 CID 56642816) is a potent and selective activator of Kir3 1/3 2 (G protein-coupled inwardly rectifying potassium GIRK1/2) channel with EC50 of 0 16 M and 1 8 M for GIRK1/2 and GIRK1/4 respectively ML-297 is potential for the treatment of epilepsy
Catalog No. 50-000-02102