Learn More
Medchemexpress LLC ML-298 10mg | 1426916-02-0 | 432.44 g·mol⁻1 | C22H23F3N4O2 | 10 MG
SDP
Catalog No. 5000226749
Encompass_Preferred
ML-298 is a potent, selective small-molecule inhibitor of phospholipase D2 (PLD2) used in biochemical and cell-based research. It inhibits PLD2 with an IC50 of 355 nM, displays strong selectivity over PLD1, and has been shown to reduce invasive migration in U87-MG glioblastoma cells.
  • Potent, selective PLD2 inhibition (IC50 355 nM).
  • High selectivity over PLD1 (IC50 ≈ 20,000 nM).
  • High purity suitable for research (≥98%).
  • Supplied in research-scale quantities for in vitro studies.
  • Stable as a powder under recommended storage conditions.
Catalog No. 50-002-26749 Supplier Medchemexpress LLC Supplier No. HY11616510MG
ML-298 is a potent, selective small-molecule inhibitor of phospholipase D2 (PLD2) used in biochemical and cell-based research. It inhibits PLD2 with an IC50 of 355 nM, displays strong selectivity over PLD1, and has been shown to reduce invasive migration in U87-MG glioblastoma cells.
  • Potent, selective PLD2 inhibition (IC50 355 nM).
  • High selectivity over PLD1 (IC50 ≈ 20,000 nM).
  • High purity suitable for research (≥98%).
  • Supplied in research-scale quantities for in vitro studies.
  • Stable as a powder under recommended storage conditions.