Learn More
TARGETMOL CHEMICALS INC ML-298 10MG
SDP
Catalog No. 502877039
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ML-298 (CID53393915) is a potent, specific inhibitor of Phospholipase D2 (PLD2, IC50 of 355 nM). Purity 99.7%
Catalog No. 50-287-7039 Supplier TARGETMOL CHEMICALS INC Supplier No. T896610MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ML-298 (CID53393915) is a potent, specific inhibitor of Phospholipase D2 (PLD2, IC50 of 355 nM). Purity 99.7%