Learn More
Cayman Chemical ML-309hydrochlorIde 10mg
SDP
Catalog No. 502987152
Encompass_Preferred
An inhibitor of mutant IDH1; reversibly and selective inhibits IDH1R132H over wild-type IDH1 (IC50s = 96 and 36,500 nM, respectively); reduces 2-hydroxyglutarate production in U87 MG glioblastoma cells (EC50 = 55 nM)
Catalog No. 50-298-7152 Supplier Cayman Chemical Supplier No. 2151910 MG
An inhibitor of mutant IDH1; reversibly and selective inhibits IDH1R132H over wild-type IDH1 (IC50s = 96 and 36,500 nM, respectively); reduces 2-hydroxyglutarate production in U87 MG glioblastoma cells (EC50 = 55 nM)