Learn More
TARGETMOL CHEMICALS INC ML-335 5MG
SDP
Catalog No. 502580902
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-335 is a u-(delta) isomer-targeted agonist a uOR-(delta)OR-biased ligand that can serve as a scaffold for the development of unique types of (isomer-biased) drugs.ML-335 is a MOR (u-opioid receptor)/DOR ((delta)-opioid receptor) heterodimer with anti-injury sensory activity and pain inhibitory activity. purity: 98%
Catalog No. 50-258-0902 Supplier TARGETMOL CHEMICALS INC Supplier No. T230025MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-335 is a u-(delta) isomer-targeted agonist a uOR-(delta)OR-biased ligand that can serve as a scaffold for the development of unique types of (isomer-biased) drugs.ML-335 is a MOR (u-opioid receptor)/DOR ((delta)-opioid receptor) heterodimer with anti-injury sensory activity and pain inhibitory activity. purity: 98%