Learn More
TARGETMOL CHEMICALS INC ML-336 5MG
SDP
Catalog No. 502879556
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.ML-336 is an inhibitor of of the Venezuelan equine encephalitis virus (VEEV) strain TC-83. Purity 97%
Catalog No. 50-287-9556 Supplier TARGETMOL CHEMICALS INC Supplier No. T219005MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.ML-336 is an inhibitor of of the Venezuelan equine encephalitis virus (VEEV) strain TC-83. Purity 97%