Learn More
Abcam ML-385, 5MG
Catalog No. 502680456
Encompass_Preferred
MW 511.6 Da, Purity >=98%. A potent and selective Nrf2 inhibitor (Nrf2; IC₅₀ = 1.9 μM). ML-385 blocks Nrf2 downstream target gene expression. ML-385 displays specificity and selectivity for NSCLC cells with KEAP1 mutation, which leads to gain of Nrf2 function.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-0456 Supplier Abcam Supplier No. AB2871095MG
MW 511.6 Da, Purity >=98%. A potent and selective Nrf2 inhibitor (Nrf2; IC₅₀ = 1.9 μM). ML-385 blocks Nrf2 downstream target gene expression. ML-385 displays specificity and selectivity for NSCLC cells with KEAP1 mutation, which leads to gain of Nrf2 function.
The product is subject to the following: Abcam Restricted Use Statement