Learn More
Medchemexpress LLC HY-122122 10mg , ML-60218 CAS:577784-91-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12212210MG
Encompass_Preferred
HY-122122 10mg ML-60218 CAS: 577784-91-9 ML-60218 is a broad-spectrum RNA pol III inhibitor, with IC50s of 32 and 27 μM for Saccharomyces cerevisiae and human. ML-60218 disrupts already assembled viroplasms and to hamper the formation of new ones without the need for de novo transcription of cellular RNAs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9323