Learn More
Selleck Chemical LLC ML-7 HCl 10mg 110448-33-4
SDP
Catalog No. 501366016
Encompass_Preferred
ML-7 is an inhibitor of smooth muscle myosin light chain kinase (MLCK) with a Ki value of 0.3 ¿M and displays reversible, ATP-competitive inhibition of Ca2+-calmodulin-dependent and -independent smooth muscle MLCKs. ML-7 also inhibits PKA and PKC with Ki of 21 ?M and 42 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6016 Supplier Selleck Chemical LLC Supplier No. S838810MG
ML-7 is an inhibitor of smooth muscle myosin light chain kinase (MLCK) with a Ki value of 0.3 ¿M and displays reversible, ATP-competitive inhibition of Ca2+-calmodulin-dependent and -independent smooth muscle MLCKs. ML-7 also inhibits PKA and PKC with Ki of 21 ?M and 42 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.