Learn More
TARGETMOL CHEMICALS INC ML-7 HYDROCHLORIDE 25MG
SDP
Catalog No. 502580412
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-7 hydrochloride (ML-7 HCl) is a cell-permeable reversible effective ATP-competitive and specific inhibitor of myosin light chain kinase (Ki 300 nM); also inhibits smooth-muscle myosin light chain kinase PKA and PKC. purity: 99%
Catalog No. 50-258-0412 Supplier TARGETMOL CHEMICALS INC Supplier No. T305025MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-7 hydrochloride (ML-7 HCl) is a cell-permeable reversible effective ATP-competitive and specific inhibitor of myosin light chain kinase (Ki 300 nM); also inhibits smooth-muscle myosin light chain kinase PKA and PKC. purity: 99%