Learn More
Medchemexpress LLC ML-792 | CAS No. : 1644342-14-2 | 10 mM * 1 mL in DMSO
SDP
Catalog No. NC2882761
Encompass_Preferred
ML-792 is a potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2 in enzymatic assays (IC50 values of 3 and 11 nM, respectively) compared with NAE/NEDD8 and UAE/ubiquitin (IC50> values of 32 μM and >100 μM, respectively).
Catalog No. NC2882761 Supplier Medchemexpress LLC Supplier No. HY10870210MM1ML DMSO
ML-792 is a potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2 in enzymatic assays (IC50 values of 3 and 11 nM, respectively) compared with NAE/NEDD8 and UAE/ubiquitin (IC50> values of 32 μM and >100 μM, respectively).