Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ML-792 | 1644342-14-2 | 99.8% | 551.41 | 50 MG
SDP

Catalog No. 5000345397
Encompass_Preferred

ML-792 is a potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2 in enzymatic assays, with IC50 values of 3 nM and 11 nM, respectively. It demonstrates significantly less activity against NAE/NEDD8 and UAE/ubiquitin. This product is for research use only.

  • Potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2.
  • IC50 values of 3 nM for SAE/SUMO1 and 11 nM for SAE/SUMO2.
  • Significantly less activity against NAE/NEDD8 and UAE/ubiquitin.
  • Purity: 99.76%.
  • CAS number: 1644342-14-2.
  • Molecular weight: 551.41.
  • Appearance: solid, white to off-white.
  • Inhibits SAE and SUMO-pathway activities in HCT116 cells.
  • Inhibits cell proliferation and decreases cancer cell viability.
  • Demonstrates dose-dependent viability effect.
  • Revealed dose-dependent decrease in SAE and UBC9 thioester levels.
  • Soluble in DMSO: 100 mg/mL (181.35 mM).

Catalog No. 50-003-45397 Supplier Medchemexpress LLC Supplier No. HY10870250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ML-792 is a potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2 in enzymatic assays, with IC50 values of 3 nM and 11 nM, respectively. It demonstrates significantly less activity against NAE/NEDD8 and UAE/ubiquitin. This product is for research use only.

  • Potent and selective inhibitor of SAE/SUMO1 and SAE/SUMO2.
  • IC50 values of 3 nM for SAE/SUMO1 and 11 nM for SAE/SUMO2.
  • Significantly less activity against NAE/NEDD8 and UAE/ubiquitin.
  • Purity: 99.76%.
  • CAS number: 1644342-14-2.
  • Molecular weight: 551.41.
  • Appearance: solid, white to off-white.
  • Inhibits SAE and SUMO-pathway activities in HCT116 cells.
  • Inhibits cell proliferation and decreases cancer cell viability.
  • Demonstrates dose-dependent viability effect.
  • Revealed dose-dependent decrease in SAE and UBC9 thioester levels.
  • Soluble in DMSO: 100 mg/mL (181.35 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.