Learn More
Cayman Chemical ML-7hydrochlorIde 10mg
SDP
Catalog No. 502954836
Encompass_Preferred
Inhibits smooth MLCK (Ki = 0.3 µM) 10-fold more potently than its parent compound ML-9
Catalog No. 50-295-4836 Supplier Cayman Chemical Supplier No. 1180110 MG
Inhibits smooth MLCK (Ki = 0.3 µM) 10-fold more potently than its parent compound ML-9