Learn More
Cayman Chemical ML-7 (hydrochloride), Purity: ≥95%, CAS: 110448-33-4, 50 mg
SDP
Catalog No. 502954173
Encompass_Preferred
Inhibits smooth MLCK (Ki = 0.3 µM) 10-fold more potently than its parent compound ML-9
Catalog No. 50-295-4173 Supplier Cayman Chemical Supplier No. 1180150 MG
Inhibits smooth MLCK (Ki = 0.3 µM) 10-fold more potently than its parent compound ML-9