Learn More
Medchemexpress LLC HY-100932 25mg Medchemexpress, ML-9 CAS:105637-50-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10093225MG
Encompass_Preferred
Medchemexpress, HY-100932 25mg ML-9 CAS:105637-50-1 ML-9 is a selective and potent inhibitor of Akt kinase, inhibits myosin light-chain kinase (MLCK) and stromal interaction molecule 1 (STIM1) activity. ML-9 inhibits inhibits MLCK, PKA and PKC activity with Ki values of 4, 32 and 54 μM, respectively. ML-9 induces autophagy by stimulating autophagosome formation and inhibiting their degradation.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0608