Learn More
Enzo Life Sciences ML-9 (50mg). CAS: 105637-50-1
SDP
Supplier:  Enzo Life Sciences BMLEI1530050
Encompass_Preferred
Selective inhibitor of MLC kinase (Ki=3.8 µM). Inhibits insulin-stimulated 2-deoxyglucose transport and PP-1 activation in adipocytes, possibly via an effect on MAP kinase or its activation. Useful PKB inhibitor (IC50=10-50 µM). Also known to inhibit PKA (IC50=20 µM) and S6 kinase (IC50=50 µM). Does not inhibit PI 3-kinase. Alternative name: 1-(5-Chloronaphthalene-1-sulfonyl)-1H-hexahydro-1,4-diazepine . HCl. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (10mg/ml) and ethanol/water (1:1, 10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-201-0574