Learn More
Medchemexpress LLC ML-9 (free base) | 110448-31-2 | 99.9% | 324.83 | C15H17ClN2O2S | 50 MG
SDP
Catalog No. 5000212592
Encompass_Preferred
ML-9 (free base) is a small-molecule kinase inhibitor used in biochemical and cell-based research to probe myosin light-chain kinase (MLCK) and Akt-related signaling. It also modulates PKA, PKC, and STIM1 activity and can induce autophagy, making it useful for studies of cytoskeleton regulation and autophagic processes.
  • Inhibits myosin light-chain kinase and Akt kinase.
  • Modulates PKA, PKC, and STIM1 activity.
  • Induces autophagy by promoting autophagosome formation.
  • Suitable for biochemical and cell-based assays.
  • Well characterized with reported chemical identifiers and molecular weight.
Catalog No. 50-002-12592 Supplier Medchemexpress LLC Supplier No. HY10093250MG
ML-9 (free base) is a small-molecule kinase inhibitor used in biochemical and cell-based research to probe myosin light-chain kinase (MLCK) and Akt-related signaling. It also modulates PKA, PKC, and STIM1 activity and can induce autophagy, making it useful for studies of cytoskeleton regulation and autophagic processes.
  • Inhibits myosin light-chain kinase and Akt kinase.
  • Modulates PKA, PKC, and STIM1 activity.
  • Induces autophagy by promoting autophagosome formation.
  • Suitable for biochemical and cell-based assays.
  • Well characterized with reported chemical identifiers and molecular weight.