Learn More
CYTOSPRING LLC ML BUFFER TRIS PH7.4 10X10ML
Catalog No. NC2249917
Encompass
T10E0.1 MI Buffer 10x10mL, 10mM Tris ph7.4, 0.1 mMEDTA in RNAse free containers (please use the 15mL tubes)
This item is not returnable. View return policy
Catalog No. NC2249917 Supplier CYTOSPRING LLC Supplier No. TE010
T10E0.1 MI Buffer 10x10mL, 10mM Tris ph7.4, 0.1 mMEDTA in RNAse free containers (please use the 15mL tubes)