Learn More
TARGETMOL CHEMICALS INC ML-SA1 25MG
SDP
Catalog No. 502580901
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-SA1 (Mucolipin synthetic agonist 1) is a selective TRPML agonist inhibits Dengue virus 2 (DENV2) and Zika virus (ZIKV) by promoting lysosomal acidification and protease activity. The IC50 value of ML-SA1 against DENV2 RNA and ZIKV RNA is 8.3 uM and 52.99 uM respectively[1]. purity: 99%
Catalog No. 50-258-0901 Supplier TARGETMOL CHEMICALS INC Supplier No. T2300425MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-SA1 (Mucolipin synthetic agonist 1) is a selective TRPML agonist inhibits Dengue virus 2 (DENV2) and Zika virus (ZIKV) by promoting lysosomal acidification and protease activity. The IC50 value of ML-SA1 against DENV2 RNA and ZIKV RNA is 8.3 uM and 52.99 uM respectively[1]. purity: 99%