Learn More
Selleck Chemical LLC ML-SA1-5mg
SDP
Supplier:  Selleck Chemical LLC S99265MG
Encompass_Preferred
ML-SA1 (Mucolipin synthetic agonist 1) is an activator of TRPML channels. ML-SA1 also inhibits Dengue virus 2 (DENV2) and Zika virus (ZIKV) by promoting lysosomal acidification and protease activity. The IC50 values of ML-SA1 against DENV2 RNA and ZIKV RNA are 8.93 μM and 52.99 μM, respectively. ML-SA1 induces autophagy. ML-SA1 can be used for the research of broad-spectrum antiviral.
Catalog No. 50-238-9960