Learn More
Abcam ML-SA1, TRPML agonist, 10MG
Catalog No. 502705555
Encompass_Preferred
MW 362.4 Da, Purity >98%. Selective TRPML agonist. Induces TRPML1, 2 and 3-mediated Ca²+ release from lysosomes in vitro. Does not activate TRPM2, TRPV2, TRPA1, TRPV3, TRPC6 channels. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5555 Supplier Abcam Supplier No. AB14462210MG
MW 362.4 Da, Purity >98%. Selective TRPML agonist. Induces TRPML1, 2 and 3-mediated Ca²+ release from lysosomes in vitro. Does not activate TRPM2, TRPV2, TRPA1, TRPV3, TRPC6 channels. Cell-permeable.
The product is subject to the following: Abcam Restricted Use Statement