Learn More
TARGETMOL CHEMICALS INC ML-SA5 10MG
SDP
Catalog No. 502576629
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-SA5 is an MCOLN1 agonist and arrests autophagic flux by perturbing fusion between autophagosomes and lysosomes. purity: 100%
Catalog No. 50-257-6629 Supplier TARGETMOL CHEMICALS INC Supplier No. T6003610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ML-SA5 is an MCOLN1 agonist and arrests autophagic flux by perturbing fusion between autophagosomes and lysosomes. purity: 100%